VWR® PerfectBlue, Tank Blotters

Leverandør: VWR Collection
700-1228EA 19925 NOK
700-1228 700-1230
VWR® PerfectBlue, Tank Blotters
Blottingapparat
PerfectBlue S and M tank electroblotters are designed to rapidly transfer nucleic acid or protein fragments from up to four polyacrylamide gels at a time to nitrocellulose, nylon or PVDF membranes. The colour coded cassettes allow for easy assembly of transfer sandwich and error-free transfer. The large buffer capacity and integral cooling allow extended transfer times and high voltage/high current operation for the transfer of high molecular weight proteins.

  • Sturdy buffer chamber for up to four colour coded blotting cartridges, with sufficient volume for
More Product Information
Bestill nå

SPESIFIKASJONER

Modell PerfectBlue S PerfectBlue M
Environmentally Preferable
Buffervolum ~1300 ml ~4500 ml
Aktivt overføringsområde 80,75 cm³ 360 cm³
Overføringstid 120 - 360 min 120 - 360 min
Enhet, B × D × H 185×145×155 mm 290×140×350 mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR