VWR® PerfectBlue, Power Supply, Universal
Leverandør: VWR Collection
700-1285EA 10100 NOK
700-1285
VWR® PerfectBlue, Power Supply, Universal
Strømkilder
VWR® PerfectBlue power supply is constructed with PC housing and flame-retardant ABS face plate, and is ideal for DNA, RNA and protein electrophoresis, as well as blotting, and for routine horizontal and vertical electrophoresis.
  • Compact size with a stackable case provides a small footprint
  • TFT-LCD (2,4") colour screen shows all parameters during operation
  • Timer with alarm function
  • Constant voltage/current/power operation mode
  • Restart the operating program automatically after power outage
The power supply is microprocessor controlled and has fourMore Product Information