Flat electrode for cell fusion
Leverandør: BTX
45-0108
700-4812EA 24475 NOK
700-4812
Flat electrode for cell fusion
Elektroder Elektroporasjonselektroder
This flat electrode can be used for both electroporation and electro cell fusion. It generates either a divergent or homogeneous field depending on the orientation of the grooved electrodes.
The flat electrode can be oriented with the grooved sides of the electrode facing one another to generate a divergent field for use in electro cell fusion. Alternatively, it can be oriented with the flat sides facing each other providing a homogeneous field for electroporation. The electrode is made of two rectangular, parallel plates of high grade stainless steel that are press-fitted into aMore Product Information