Temperature measuring for urine
Leverandør: SURESCREEN
TEMP
700-9660EA 580 NOK
700-9660
Temperature measuring for urine
Narkotester
For use with the standard collection cup, and used to show if a urine sample is within range of body temperature.
  • Samples outside this temperature range could have been adulterated