Rhodamine 123 mitochondria dye, for fluorescence microscopy

Leverandør: Biotium
Warning

Synonymer: 3,6-Diamino-9-(2-(methoxycarbonyl)phenyl xanthylium chloride, 3,6-Diamino-9-(2-(methoxycarbonyl)phenyl)xanthylium chloride, 2-(6-Amino-3-imino-3H-xanthen-9-yl)benzoic acid methyl ester

70010
70010.EA 2125 NOK
70010.
Rhodamine 123 mitochondria dye, for fluorescence microscopy
Rhodamine 123

Rhodamine 123 is a popular green fluorescent mitochondrial dye that stains mitochondria in living cells in a membrane potential-dependent fashion. It is widely used in flow cytometry studies involving mitochondrial membrane potential.


  • Cellular localization: Mitochondria
  • For live or fixed cells: For live/intact cells
  • Assay type/options: Real-time imaging
  • Cell permeability: Membrane permeant
  • Apoptosis/viability marker: Mitochondrial potential
  • Potential dependence: Mitochondrial potential-dependent
  • Colors: Red
  • Excitation/Emission: 505/534 nm
  • Orange red solid soluble in DMSO or DMF
  • Store at 4°C and protect from light, especially in solution


Please also see Dihydrorhodamine 123 and its HCl salt (10055 and 10056).

Formel: C₂₁H₁₇ClN₂O₃
MW: 380.82 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00012664
CAS-nummer: 62669-70-9

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR