behrotest® DET supplementary set for Soxhlet extraction (100 ml) DE 100
Leverandør: BEHR
B00218118
701-0069EA 41725 NOK
701-0069
behrotest® DET supplementary set for Soxhlet extraction (100 ml) DE 100
Ekstraktorer Soxhlet- og Soxhlet-typeuttrekkere
Soxhlet extraction designed for larger solid samples with five sample positions.
This set includes the following equipment:
5×Sample vessels, 250 ml 65/250
5×Vessel –extractor connector M 2965
5×100 ml soxhlet extractor EZ 100
5×Reflux condenser for EZ 100 extractor RFK 100
Bestillingsinformasjon: The DET Basic unit (DBAS, 705-0023) is also required.