Groundwater sampling capsules, Polycap™ GW, Whatman™
Leverandør: Whatman products (Cytiva)
6724-6004 6714-6004
701-0173EA 115100 NOK
701-0173 701-0164
Groundwater sampling capsules, Polycap™ GW, Whatman™
Filter Membranfilterkapsler
These Polycap™ GW (Ground Water) capsule filters are specifically designed for sample preparation of ground water samples for dissolved metals analysis. The in-line design with stepped hose barbs and hydrophilic polyethersulfone (PES) membrane make it easy to use for field sampling.
  • Stepped hose barb fittings allow for connection directly to outlet of a sampling pump with various size tubing
  • Effective filtration surface area of 600 cm² to ensure rapid sample collection
  • PES filters in thermally fused housing eliminate sample contamination that could come
More Product Information
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Vanngjennomstrømning 60 l/min (1,0 bar)
Filtreringsområde 600 cm²
Inngang/utgang 6 to 9 mm stepped barb
Maks. trykk 4,1 bar