Automatic Kjeldahl distillation unit, UDK 149
Leverandør: VELP SCIENTIFIC
F30200140
701-0225EA 0 NOK
701-0225
Automatic Kjeldahl distillation unit, UDK 149
Destillasjonssystemer Kjeldahl-destillasjonssystemer
The UDK 149 is a more flexible solution for laboratories performing Kjeldahl distillation. It is fully automatic and can be easily connected to a large choice of external titrators. The unit operates automatically, after setting water, boric acid and sodium hydroxide addition, distillation time and the steam generation output level between 10 and 100%. Different automatic titrator models can be connected for direct output of the final result. The high precision pumps ensure constant accurate dosing of reagents.
  • Automatic NaOH, H₂O and H₃BO₃ addition
  • Delay time (Devarda alloy analysis)