Sodium-2-naphthalenesulphonate ≥99.0% (by HPLC) for ioneparkromatografi, Supelco®

Leverandør: Merck


M-Clarity™ quality level = MQ100

70289-10G
70289-10GEA 2040 NOK
70289-10G
Sodium-2-naphthalenesulphonate ≥99.0% (by HPLC) for ioneparkromatografi, Supelco®
Sodium-2-naphthalenesulphonate
Formel: C₁₀H₈O₃S
MDL nummer: MFCD00064186
CAS-nummer: 532-02-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6603c8710e84184a6780a608c085dd4d2b530b69&vid=cab8608828970c894f3d0620668082de2789cfed&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR