Natriumhydroksid 2 mol/l (2 N) in aqueous solution, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®

Leverandør: Avantor
Danger

Synonymer: Caustic soda

7036.1000EA 485 NOK
7036.1000
Natriumhydroksid 2 mol/l (2 N) in aqueous solution, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Natriumhydroksid
Formel: NaOH
Kokepunkt: 109 °C (1013 hPa)
Tetthet: ≤1,11 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003548
CAS-nummer: 1310-73-2
EINECS: 215-185-5
FN: 1824
ADR: 8,II

Bestill nå

Spesifikasjon Testresultater

Titer (mol/l) 1.995 - 2.005
Trace Impurities (in ppm)
Chloride (Cl) max. 5
Iron (Fe) max. 0.5
Heavy Metals (as Pb) max. 1

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR