behrotest® DET supplementary set for Soxhlet extraction (30 ml), DE 30
Leverandør: BEHR
B00218117
705-0026EA 34250 NOK
705-0026
behrotest® DET supplementary set for Soxhlet extraction (30 ml), DE 30
Ekstraktorer Soxhlet- og Soxhlet-typeuttrekkere
This set includes the following equipment:
5×Sample vessels, 250 ml 65/250
5×Vessel –extractor connector M 2965
5×30 ml soxhlet extractor EZ 30
5×Reflux condenser for EZ 30 extractor RFK 30
Bestillingsinformasjon: The DET Basic unit (DBAS, 705-0023) is also required.