EDTA dinatrium salt dihydrat

Leverandør: Hach

20426
705-0079EA 445 NOK
705-0079
EDTA dinatrium salt dihydrat
EDTA dinatrium salt dihydrat
Formel: C₁₀H₁₄N₂Na₂O₈·2H₂O
Smeltepunkt: 255 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00150037
CAS-nummer: 6381-92-6
EINECS: 205-358-3
Merck Indeks: 13,03543
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f89002087c5d4d84af5dded62c87287b1774fab&vid=0ef4c3febbcc8a20569606067799db31f2540748&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR