Test kits, photometric, AccuVac®
Leverandør: Hach
2502025 2503025 2507025 2515025 2508025 2516025 2517025 2518025 2501025
705-0146EA 630 NOK
705-0146 705-0147 HACH2507025 705-0148 HACH2508025 705-0149 705-0150 705-0151 705-0145
Test kits, photometric, AccuVac®
Miljø testkit
AV - AccuVac® - Sealed glass cuvettes containing a portioned amount of reagent. Simply immerse the tip of the accuvac in the sample, break it and the vacuum draws the sample into the cuvette.