Digestion system, HT 200S
Leverandør: Hach
LTV077
705-0561EA 47675 NOK
705-0561
Digestion system, HT 200S
Destruksjonssystemer Arbeidsstasjoner for Mineraliseirng
This digestion system has high speed digestion technology for fast and cost effective digestion in 15 min.
  • Saves time in the analysis of COD, TNb, Ptot and heavy metals
  • Automatic fast cooling
  • COD results in 35 min
  • Variable digestion time and temperature for special digestions