Klorinanalysatorer, ChloroSense®
Leverandør: Palintest
CS100
705-0650EA 26025 NOK
705-0650
Klorinanalysatorer, ChloroSense®
Analysatorer Elementanalysatorer
ChloroSense er en reagensfri, operatøruavhengig metode for bestemmelse av fritt og totalt klorin på mindre enn 1 minutt.
  • Alternative to the DPD method ((N,N-diethylparaphenylenediamine)
  • Reagent-free and operator independent method
  • Patented sensor for fast and accurate results
Designed for drinking water, it is used in other applications including washing of food produce, testing of potable water storage tanks, pipe refurbishment as well as in hospitals.
Leveringsinformasjon: Leveres med 100 elektroder, kalibreringsbrikke, prøveflasker og myk bæreveske.