Handheld turbidity meters, Turb® 430
Leverandør: WTW
600320 600325 600326 600321
705-1027EA 40750 NOK
705-1027 662-2147 705-0672 705-0671
Handheld turbidity meters, Turb® 430
Turbiditetsmålere
The Turb 430, with lab option via LabStation, include drinking water testing, environmental monitoring, laboratory reference measurements and process control.
  • Nephelometric measurements at 90° scattered light
  • Optional sets include rugged carrying case for field use
  • LabStation LSFlex for easy lab use as optional accessories
Sertifiseringer: Scattered light characteristics according to the European Pharmacopoeia 5,0.
AQA for data evaluation by interval setting and GLP compliant data export to a PC with LS data software option.
Leveringsinformasjon: More Product Information