Turbidity sensor for MultiLine® IDS meters, VisoTurb® 900-P
Leverandør: WTW
600700
705-0917EA 33225 NOK
705-0917
Turbidity sensor for MultiLine® IDS meters, VisoTurb® 900-P
Elektroder Elektroder, øvrige
VisoTurb® 900-P is an IR turbidity sensor for applications where turbidity can be directly measured in the sample. Its application range includes measurements in surface water, pumping tests and effluents of wastewater treatment plants and other spot measurements.
  • Robust and reliable with exclusive corrosion-proof titanium housing, waterproof and pressure resistant up to 10 bars
  • Works with all Multiline® IDS handheld meters, also in combination with additional IDS sensors (2- and 3-channel meters)
  • Wireless ready: Works with IR for mobile applications
  • Can