Caps for culture tubes
Supplier: Hach
2241106
705-1201EA 80 NOK
705-1201
Caps for culture tubes
Lokk Lokk til reagensglass
Screw cap with fluoropolymer liner.
  • Firmly bonded liner provides a highly inert sealing surface
Order Now