Flocculators
Leverandør: VELP SCIENTIFIC
F105A0108 F105A0109 F105A0112 F105A0111 F105A0117
705-1501EA 20800 NOK
705-1501 705-1502 705-1504 705-1503 705-1500
Flocculators
Flokktester
The FP4, FC S, and JLT Series assist laboratory technicians in the environmental sector by providing tools for conducting the jar test and leaching test, which are essential for water treatment and environmental protection. These tests help determine the most effective methods for improving water quality and assessing the potential environmental risks posed by pollutants.
  • Timer function for FP4 and JLT units
  • Stainless steel stirring rods are adjustable in height
  • Strong metallic frame, epoxy coated for protection against chemical and mechanical corrosion