J.T.Baker® BAKERBOND®, Florisil Bulk Packing Material
Leverandør: Avantor
7061-00EA 735 NOK
7061-00
J.T.Baker® BAKERBOND®, Florisil Bulk Packing Material
Kromatografiabsorbenter SPE sorbenter
SPE sorbent for clean-up columns. Suitable for the determination of Hydrocarbon Oil Index according ISO 9377-2 and NEN 5733.