Natriumhydroksid 97.0-100.5% Ph. Eur., Macron Fine Chemicals™

Leverandør: Avantor Performance Materials

7066-33EA 355 NOK
7066-33 7066-36 7066-38
Natriumhydroksid 97.0-100.5% Ph. Eur., Macron Fine Chemicals™
Natriumhydroksid
Formel: NaOH
MDL nummer: MFCD00003548
CAS-nummer: 1310-73-2
FN: 1823
ADR: 8,II

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR