Kaliumperoksodisulfat ≥99.0%, AR® ACS, Macron Fine Chemicals™

Leverandør: Avantor Performance Materials
Danger

7076-20EA 0 NOK
7076-20
Kaliumperoksodisulfat ≥99.0%, AR® ACS, Macron Fine Chemicals™
Kaliumperoksodisulfat
Formel: K₂S₂O₈
Smeltepunkt: 100 °C
Tetthet: 2,477 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011386
CAS-nummer: 7727-21-1
EINECS: 231-781-8
FN: 1492
ADR: 5.1,III
Merck Indeks: 13,07741

Bestill nå

Spesifikasjon Testresultater

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use = 99.0 %
ACS - Assay (K₂S₂O₈) ≥99.0 %
ACS - Chlorine Compounds (as Cl) ≤0.001 %
ACS - Heavy Metals (as Pb) ≤0.001 %
ACS - Insoluble Matter ≤0.005 %
ACS - Iron (Fe) ≤0.0005 %
ACS - Manganese (Mn) ≤0.0002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR