AutoKjel Autosampler for VELP UDK 169 Automatic Kjeldahl Nitrogen Protein Analyser
Leverandør: VELP SCIENTIFIC
F30200430
709-0140EA 0 NOK
709-0140
AutoKjel Autosampler for VELP UDK 169 Automatic Kjeldahl Nitrogen Protein Analyser
Autosamplere
The UDK 169 and AutoKjel Kjeldahl analyser has been developed for continuous operation, providing the maximum sample throughput and data processing automation.
  • Developed for continuous operation
  • Provides maximum sample throughput and data processing automation
The UDK 169 used in combination with the AutoKjel autosampler, for a highly productive system, is capable of autonomously processing up to 24 samples with standard test tubes (250 ml) or 21 Jumbo test tubes (400 ml). Sample tubes are taken from the carousel and fed directly into UDK 169 forMore Product Information