Transfer pipettes, Transpette™, SP Bel-Art
Leverandør: Bel-Art Products, a Part of SP
F378730001
710-0083EA 9975 NOK
710-0083
Transfer pipettes, Transpette™, SP Bel-Art
Pipetter Pasteurpipetter og overføringspipetter
Economical, single-use, 8-channel transfer pipette made of translucent plastic. Ideal for washing plates, feeding and supernatant aspiration from microwell plates.
  • No expensive pipettes and pipette tips needed
  • Should be disposed directly after 'hot' uses in order to avoid radiation contamination of costly pipettes
  • Available sterile or non sterile; can't be steam sterilised
  • Capacity: 600 µl per channel