Colony counters
Leverandør: IUL Instruments SA
90008701
710-0106EA 17850 NOK
710-0106
Colony counters
Kolonitellere
Choice of models with a fibre marker with a built-in switch and /or a metal-tipped pen that are attached to the base. The marker supplied with the counter allows marked cells to be added to the screen one by one.
  • Ø 100 or 120 mm biconvex lamp
  • X2 magnification
220 V voltage, 50 Hz, 0,2 A.