Colony counter, Haloes Caliper
Leverandør: IUL Instruments SA
9000900
710-0112EA 46500 NOK
710-0112
Colony counter, Haloes Caliper
Kolonitellere
The Haloes Calliper is an designed to measure the diameter of haloes or objects in fluid culture media in Petri dishes, such as for antibiotic testing.
  • High quality lighting system
  • Measures without contact
The lens of the device illuminates the Petri dish and provides a magnified image of the object on a calibrated grid, divided into mm.