VWR® M40, Transportvattpinne
Leverandør: VWR Collection
710-0442EA 735 NOK
710-0442
VWR® M40, Transportvattpinne
Svabere Svabere for transport
Transport swab with plastic shaft, synthetic tip, and Amies agar gel.
  • Compliant with CLSI (formerly NCCLS) standard M40-A at 21 and 4 °C
  • Without charcoal
Helps to improve recovery and survival of fastidious microorganisms from the throat, vagina, wounds and skin swabs.
Sertifiseringer: CLSI (formerly NCCLS).
NB: *Produktet/produktene på denne siden er utenfor omfanget av EU -direktivet 2019/904