Svaber, medium CleanFoam®-hode
Leverandør: Texwipe
TX740B
710-0555EA 13725 NOK
710-0555
Svaber, medium CleanFoam®-hode
Svabere Svabere for rengjøring
Swab with a thermally-moulded CleanFoam head on a flexible paddle. Made from PU foam with 100 pore per inch porosity.
  • Very low in both non volatile residue and particle generation
  • Good chemical resistance
  • Long handle and flexible internal head paddle for support
Designed for industrial applications from general assembly to cleanrooms with a solvent such as isopropanol.
Forpakning: Bag-within-a-bag packaging.
NB: *The product(s) on this page are outside the scope of the EU Directive 2019/904