Svaber, Micro CleanFoam®
Leverandør: Texwipe
TX757B
710-0578EA 1975 NOK
710-0578
Svaber, Micro CleanFoam®
Svabere Svabere for rengjøring
PU foam head has been solvent-washed to greatly reduce non volatile residue.
  • Very low particle generation
  • Swab incorporates a flexible paddle
  • Excellent sorbency and good chemical resistance
Excellent swab for cleaning very small areas of parts and assemblies with solvents such as IPA, applying lubricants and other liquids, and removing excess materials.
Forpakning: Renromsbehandlet. Pakket pose i pose.
NB: *Produktet/produktene på denne siden er utenfor omfanget av EU -direktivet 2019/904