Gravimetric dilutor, IUL pinch Dilutor
Leverandør: IUL Instruments SA
7511
710-0862EA 26875 NOK
710-0862
Gravimetric dilutor, IUL pinch Dilutor
Dilutere Gravimetriske dilutere
Equipment specifically designed to facilitate the controlled addition of media into a homogeniser/lab blender bag using a load cell. The Pinch Dilutor constantly monitors the weight in the homogeniser bag and consequently distributes the precise quantity of diluent into the bag.
  • Ideal for dilution of solid and semi-solid samples
  • Simple to operate and control dilution with pinch valve
  • Smooth, silent and fast process
  • Contact parts and tubing can easily be dismantled and sterilised in an autoclave