VWR®, Engangshoder for luftprøvetakere
Leverandør: VWR Collection
710-0890EA 2575 NOK
710-0890 710-0891
VWR®, Engangshoder for luftprøvetakere
Prøvetakere Luftprøvetakere
Environmental monitoring in cleanrooms is important and a critical part of the production chain for a manufacturing company.
  • Made of 100% recyclable plastic
  • Ready to use head for quick actions and emergency situations
  • Can enter the sterile cleanrooms without contamination issues
Sertifiseringer: cGMP.
Bestillingsinformasjon: Includes a certification of sterilisation.
Forpakning: Double-wrap ensures complete sterility of the head while entering a cleanroom and a long shelf life (5 years).