Spiral plater, WASP 2
Leverandør: DON WHITLEY SCIENTIFIC
S00600
710-0915EA 0 NOK
710-0915
Spiral plater, WASP 2
Spiralutstrykere
Spiral platers are used extensively in the food industry to test for safe levels of bacteria; non-food applications include pharmaceuticals, cosmetics, water, and mouth and skin bacteria. The WASP 2 (Whitley Automatic Spiral Plater) takes advantage of contemporary technology to offer microprocessor-controlled stepper motor motion to streamline and automate traditional spiral plate techniques.
  • Eliminates the need for serial dilutions
  • Requires minimal set up time
  • Reduces cost per test
  • Standardises counting methods
Leveringsinformasjon: Includes waterMore Product Information