VWR®, Hand Tally Counter
Leverandør: VWR Collection
710-0935EA 665 NOK
710-0935
VWR®, Hand Tally Counter
Celletellere Håndteller
Metal construction is designed for reliability, long life and hard use. Unique engineering provides a smooth tactile feel. Numbers instantly and automatically return to zero with a twist of the reset knob.
  • Compact unit for carrying in the pocket
  • Ease in viewing, large white numerals on a black background
Hundreds of laboratory applications provide opportunities to count blood samples, take surveys, inventory items, total drops of liquid, receive lab samples, and tally biomedical events. A protective window seals out dust, dirt and humidity. The rugged,More Product Information