VWR®, SAS Super Isolator luftprøvetakere
Leverandør: VWR Collection
710-0943EA 47625 NOK
710-0943 710-0944 710-0977 710-0978 710-0945 710-0946
VWR®, SAS Super Isolator luftprøvetakere
Prøvetakere Luftprøvetakere
The SAS instrument control unit is placed outside and connected to the aspirating head using a simple electrical connection. All possible typical contamination sources from the vacuum sampling system are avoided using the SAS Super Isolator. An interesting application is that several heads can be controlled by a single SAS control unit, providing a practical and economical solution.
  • Operational flexibility, multiple language choice
  • Records day, time, site, volume of air sampled and operator's name
  • Large backlit screen with intuitive menu; active operation
More Product Information
Bestill nå
SPESIFIKASJONER
Modell SAS Super Isolator 100 SAS Super Isolator 180 Duo SAS Super 360 Isolator
Environmentally Preferable
Gjennomstrømningshastighet 100 L/min 180 L/min 2×180 L/min
Sampling head Stainless steel Stainless steel Stainless steel
W×D×H 105×110×290 mm 105×110×290 mm 360×160×180 mm
Weight 1,75 kg 1,75 kg 2,1 kg