VWR®, SAS Super Pinocchio II luftprøvetaker for komprimerte gasser
Leverandør: VWR Collection
710-0950EA 56825 NOK
710-0950 710-0949 710-0955 710-0951
VWR®, SAS Super Pinocchio II luftprøvetaker for komprimerte gasser
Prøvetakere Luftprøvetakere
SAS Super Pinocchio II is a microbial impaction sampler to test the quality of compressed air and gases used in cleanrooms. A compressed air source is connected and the flow meter regulated to achieve the required flow rate.
  • Autoclavable
  • Calibrated according to International Standards
  • Unit requires no power and is fully transportable
  • Can collect air samples on either contact plates or 90 mm petri dishes
Sertifiseringer: IQ, OQ and PQ documentation available.
Leveringsinformasjon: Self-contained unit, consisting of connections, airflow meter,More Product Information