Coliform swab
Leverandør: HYSERVE
1000101
710-1479EA 2550 NOK
710-1479
Coliform swab
Svabere Svabere for prøvetaking
Easy to carry early warning test for contamination of surfaces with coliform bacteria. Based on a certified method, the Coliform Swab provides a simple method for the qualitative detection of these bacteria - with a clear colour change from purple to yellow in the presence of coliform bacteria.
  • Simple colour change shows contamination, no colour change indicates the corresponding contamination
  • Individually packed straightening test, which can be carried out without additional reagents
  • Results can be read within 18 hours
  • Determines the number of germs from 1