Media preparator, ProfiClave
Leverandør: BioTool
620001
710-1670EA 0 NOK
710-1670
Media preparator, ProfiClave
Mediefremstillinger
The media preparators ProfiClave PC10B/20B prepare up to 10/15 L of plain or complex vitamin/blood added agar formulations easier and faster than ever. Equipped with intuitive software, accessible via a convenient touch screen, training on how to use the media preparator is short and easy. ProfiClave PC20 prepares between 1 to 15 L, ProfiClave PC10 prepares between 0,5 to 10 L of agar, stirred and temperature controlled. Vessel access is simple, no bolts or tools required. Enhanced user programming capabilities (3 * 20 programs), operating parameter print-out (integrated in front panel),More Product Information