Spiral plater, WASP
Leverandør: DON WHITLEY SCIENTIFIC
S02600
710-1961EA 0 NOK
710-1961
Spiral plater, WASP
Spiralutstrykere
The Whitley automated spiral plater (WASP) eliminates serial dilutions. It offers a real reduction in the cost per test and standardises counting methods. It is used in the food industry to test for safe levels of bacteria and as a quality assurance and research tool in pharmaceuticals, water and dental practices.
  • Use without any consumables ensuring minimal running costs
  • A single keystroke operation loads the sample, inoculates a plate and cleans the stylus to optimise sample throughput speed
  • It can dispense volumes from 10 µl to 400 µl providing immense