VWR® Auto lite, Automatic Colony Counters
Leverandør: VWR Collection
710-2072EA 60325 NOK
710-2072
VWR® Auto lite, Automatic Colony Counters
Kolonitellere
Auto lite is the latest affordable, automated colony counting device. It is available in one configuration and features simple to use software that greatly improves efficiency, reliability and productivity. Auto lite software uses a powerful algorithm for colony counting on pour and spiral plates. A colony separator automatically splits touching colonies to further improve the accuracy of the counting process.
  • Low cost entry into electronic colony counting, pour and spiral
  • Results and plate images are saved to a PC enabling traceability, consistency
More Product Information
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Count Separation of touching colonies, exclusion areas
Belysning Multi-array LED lighting (computer controlled), lower lighting with upper reflective lighting for all applications
Kamera Integrated, sensitive 1 MP
Oppløsning For standard 90 mm Petri dish, smallest detectable colony is 100 - 120 μm
Programvare Windows 7/10 compatible
B×D×H 260×260×260 mm
Weight 3,3 kg