VWR®, SAS Super ISO luftprøvetakere
Leverandør: VWR Collection
710-2087EA 0 NOK
710-2087 710-2088 710-2089 710-2090
VWR®, SAS Super ISO luftprøvetakere
Prøvetakere Luftprøvetakere
SAS Super ISO 100 and SAS Super ISO 180 are differentiated by their airflow; this choice depends on the application, and allows continuous or sequential sampling from a few minutes to several hours. The data can be stored on the SAS instrument itself or on any remote PC.
  • Design avoids turbulence in unidirectional airflow and re-aspiration of tested air
  • Portable and easily positioned, they operate with long life, rechargeable batteries
  • All manuals are available on a USB key in six languages
  • Micro USB for data download and instrument configuration
  • New Data
More Product Information
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Sampling head Aluminium, stainless steel or plastic (certified ready to use)
W×D×H 120×125×275 mm
Weight 1,8 kg