VWR®, Gravimetric Dilutor, 3 kg
Leverandør: VWR Collection
710-2092EA 0 NOK
710-2092
VWR®, Gravimetric Dilutor, 3 kg
Dilutere Gravimetriske dilutere
Gravimetric dilutors are used for the dilution of a sample before microbiological analysis.
  • Available with one or two pumps
  • All stainless steel with an innovative design
  • Ultra-low profile for easy use under laminar flow
  • USB port
  • Dilution time for a 25 g sample diluted at 1/10 in less than 9 seconds
  • <1% of sample weighing
  • As little as <1% of dispensing (in standard or accurate mode)
  • The bag is maintained with the bag holder
Easy to use, with a removable drip tray to make cleaning easy, it is an accurate device in compliance with the latestMore Product Information