VWR®, Peristaltic Pump, Media

Leverandør: VWR Collection
710-2126EA 46075 NOK
710-2126
VWR®, Peristaltic Pump, Media
Pumper Slangepumper
Peristaltic pump for culture media preparation. Use it for serial dispensing of culture media, agar or any liquid diluent for efficient sample preparation.

  • Select from three dispensing modes: Continuous, dose (pre-determined volume), multi-dose (pre-determined volume, with an adjustable pause between each dose)
  • Stainless steel and heavy-duty polycarbonate top shell
  • Rotating button for easy navigation
  • Compact and ergonomic: Only 160 mm wide
  • Typical precision with inner Ø 3,2 mm tubing: 9 ml: ±1,8%, 18 ml: ±1%, 225 ml: ±0,5%
  • Dispenses liquid from 50 μl - 99
More Product Information
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR