VWR® BagMixer® 400, Laboratory Blenders
Leverandør: VWR Collection
710-2143EA 0 NOK
710-2143
VWR® BagMixer® 400, Laboratory Blenders
Homogenisatorer Labblendere
The VWR® Star Blender 400 Silent is a highly efficient laboratory blender for solid sample preparation. With a sterile bag, it guarantees optimal bacterial extraction in less than 60 seconds, and eliminates all risks of cross contamination.
  • Door opens at 270° and paddles stop side be side at end of blending for easy bag extraction
  • Removable paddles and stainless steel construction for ease of cleaning
  • Easy control of blending time, power and speed with VW CC, S and SW models
  • Q-Tight®, special closing system prevents risk of liquid leaking from the top of