VWR®, Media Preparator Premium 15 MPP15
Leverandør: VWR Collection
710-2220EA 0 NOK
710-2220
VWR®, Media Preparator Premium 15 MPP15
Mediefremstillinger
This medium preparator is equipped with everything that simplifies the production of culture media and accelerates the process. The medium is effectively mixed, heated, sterilised and quickly cooled to the desired filling temperature right from the start. For the filling process, the medium is kept stable at the selected filling temperature.
  • Designed for ease of use, the MPP15 is controlled via a large integral screen using a suite of powerful icon-based software that intuitively guides you through the culture media preparation process with on-screen