VWR® MCC 2000 Manual Colony Counter
Leverandør: VWR Collection
710-2246EA 18325 NOK
710-2246
VWR® MCC 2000 Manual Colony Counter
Kolonitellere
The MCC 2000 is designed for the rapid and accurate counting of bacterial colonies.
The MCC 2000 colony counter is an easy-to-use instrument for precise working with Petri dishes of various dimensions. The surface is sensitive to touch, therefore every single mark with the pen is additionally indicated with sound and number. The controller is equipped with acoustic counting control and automatic compensation of the weight of different Petri dishes. Even illumination of the counting field is guaranteed by a ring light. The magnifying glass ensures a clear view. Its position can be easilyMore Product Information