ReadyTube™12 MSRV Medium ISO 6579
Leverandør: MERCK LIFE SCIENCE
1466220100
710-2265EA 3960 NOK
710-2265
ReadyTube™12 MSRV Medium ISO 6579
Mikrobiologiske medier Media, preparerte for mikrobiologi
  • Application: Selective enrichment for detection of Salmonella
  • Packaging: 12 ml in 17 ml tube with red cap (100 tubes per box)
  • Synonym: Modified semi-solid Rappaport-Vassiliadis Medium