VWR® MediaBag TW Dehydrated Culture Media in Bags
Leverandør: VWR Chemicals
7101.00450EA 9225 NOK
7101.00450
VWR® MediaBag TW Dehydrated Culture Media in Bags
Mikrobiologiske medier Media, preparerte for mikrobiologi
MediaBags TW (Twirl-bag) are filled with pre-weighed granulated media, double-bagged bag and irradiated. Add sterile water under aseptic conditions.