Natriumtiosulfat 0.1 mol/l (0.1 N) vandig løsning, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®

Leverandør: Avantor

7101.1000EA 1125 NOK
7101.1000 7101.9010 7101.9020
Natriumtiosulfat 0.1 mol/l (0.1 N) vandig løsning, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Natriumtiosulfat
Formel: Na₂S₂O₃
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003499
CAS-nummer: 7772-98-7
EINECS: 231-867-5

Bestill nå

Spesifikasjon Testresultater

Titer (mol/l) 0.0997 - 0.1003
Trace Impurities (in ppm)
Heavy Metals (as Pb) max. 1
Sulfate and Sulfite (as SO4) max. 50


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR