Svovelsyre 0.05 mol/l (0.1 N) vandig løsning, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®

Leverandør: Avantor

7103.1000EA 895 NOK
7103.1000 7103.9010 7103.9020
Svovelsyre 0.05 mol/l (0.1 N) vandig løsning, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Svovelsyre
Formel: H₂SO₄
Tetthet: 1,01 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00064589
CAS-nummer: 7664-93-9
EINECS: 231-639-5
FN: 2796
ADR: 8,II

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR