Svovelsyre 0.25 mol/l (0.5 N) in aqueous solution, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®

Leverandør: Avantor

7110.1000EA 445 NOK
7110.1000
Svovelsyre 0.25 mol/l (0.5 N) in aqueous solution, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Svovelsyre
Formel: H₂SO₄
MW: 98.08 g/mol
Kokepunkt: 330
Smeltepunkt: 10.38
Tetthet: 1.01 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00064589
CAS-nummer: 7664-93-9
EINECS: 231-639-5
FN: 2796
ADR: 8 II

Bestill nå

Spesifikasjon Testresultater

Titer (N) 0.4975 - 0.5025
Trace Impurities (in ppm)
Chloride (Cl) max. 1
Iron (Fe) max. 0.5
Heavy Metals (as Pb) max. 0.3
Phosphate (PO4) max. 1


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR